Learn More
Medchemexpress LLC Benzyl-PEG5-amine | 86770-77-6 | 98.0% | C17H29NO5 | 250 MG
SDP
Catalog No. 5000352741
Encompass_Preferred
Benzyl-PEG5-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. They leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Used in PROTAC synthesis
  • PEG-based linker
  • Facilitates selective protein degradation
Catalog No. 50-003-52741 Supplier Medchemexpress LLC Supplier No. 250MGHY140743
Benzyl-PEG5-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein intended for degradation. They leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Used in PROTAC synthesis
  • PEG-based linker
  • Facilitates selective protein degradation