Learn More
Medchemexpress LLC Benzyl-PEG5-amine | 86770-77-6 | 98.0% | C17H29NO5 | 50 MG
SDP
Catalog No. 5000356637
Encompass_Preferred
Benzyl-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56637 Supplier Medchemexpress LLC Supplier No. HY14074350MG
Benzyl-PEG5-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins