Learn More
Medchemexpress LLC Benzyl-PEG5-amine | 86770-77-6 | MFCD28976656 | 98.0% | C17H29NO5 | 100 MG
SDP
Catalog No. 5000353357
Encompass_Preferred
Benzyl-PEG5-amine is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds characterized by two distinct ligands joined by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. This design enables PROTACs to utilize the intracellular ubiquitin-proteasome system to achieve the selective degradation of specific target proteins.
  • Functions as a PROTAC linker.
  • Supports the synthesis of PROTACs.
  • Engages the ubiquitin-proteasome system for protein degradation.
  • Enables selective degradation of target proteins.
Catalog No. 50-003-53357 Supplier Medchemexpress LLC Supplier No. HY140743100MG
Benzyl-PEG5-amine is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs are compounds characterized by two distinct ligands joined by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. This design enables PROTACs to utilize the intracellular ubiquitin-proteasome system to achieve the selective degradation of specific target proteins.
  • Functions as a PROTAC linker.
  • Supports the synthesis of PROTACs.
  • Engages the ubiquitin-proteasome system for protein degradation.
  • Enables selective degradation of target proteins.