Learn More
Medchemexpress LLC Benzyl-PEG7-amine 5mg
SDP
Catalog No. 5000204700
Encompass_Preferred
Benzyl-PEG7-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04700 Supplier Medchemexpress LLC Supplier No. HYW0960775MG
Benzyl-PEG7-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]