Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl piperazine-1-carboxylate | 31166-44-6 | 220.27 | 25 G
SDP

Catalog No. 5000448119
Encompass_Preferred

Benzyl piperazine-1-carboxylate is a synthetic intermediate useful in the research of melanocortin-4 (MC4) receptor antagonists. It can be used to generate high-affinity mono-piperazine MC4 receptor antagonists. This product is for research use only.

  • Synthetic intermediate for MC4 receptor antagonists
  • Generates high-affinity mono-piperazine MC4 receptor antagonists
  • Appears as a colorless to off-white solid-liquid mixture
  • Recommended storage at 4°C, protected from light
  • Soluble in DMSO at 200 mg/mL

Catalog No. 50-004-48119 Supplier Medchemexpress LLC Supplier No. 25GHYW002603
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl piperazine-1-carboxylate is a synthetic intermediate useful in the research of melanocortin-4 (MC4) receptor antagonists. It can be used to generate high-affinity mono-piperazine MC4 receptor antagonists. This product is for research use only.

  • Synthetic intermediate for MC4 receptor antagonists
  • Generates high-affinity mono-piperazine MC4 receptor antagonists
  • Appears as a colorless to off-white solid-liquid mixture
  • Recommended storage at 4°C, protected from light
  • Soluble in DMSO at 200 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.