Learn More
Medchemexpress LLC Benzyl-piperazine-co-benzothiazole-4-methylpiperidine | 906258-48-8 | 99.0% | C25H30N4OS | 10MG

Supplier: Medchemexpress LLC HY14575510MG
Benzyl-piperazine-CO-benzothiazole-4-methylpiperidine is a research-grade small molecule reported to alter the lifespan of a eukaryotic organism. Supplied as a high-purity solid for laboratory studies, it is intended for research use only and requires refrigerated or cryogenic storage for long-term stability.
- High purity (99.0%) suitable for analytical and biological studies.
- Molecular formula C25H30N4OS and molecular weight 434.60 g/mol.
- White to off-white solid, convenient for weighing and dissolution.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Provided with supporting documentation such as datasheet, COA, and SDS.
- For research use only; not for human or clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.