Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl propionate, 99%, Thermo Scientific™

Catalog No. AAB2318630
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2318630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2318630 Supplier Thermo Scientific Chemicals Supplier No. B2318630

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-63-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00027003
InChI Key VHOMAPWVLKRQAZ-UHFFFAOYSA-N
Synonym benzyl propionate, propanoic acid, phenylmethyl ester, propionic acid, benzyl ester, phenylmethyl propanoate, propionic acid benzyl ester, phenylmethyl propionate, benzyl propionate natrual, benzyl n-propionate, fema no. 2150, propionic acid, benzyl ester 6ci,7ci,8ci
PubChem CID 31219
IUPAC Name benzyl propanoate
SMILES CCC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 221°C to 223°C
Density 1.027
Boiling Point 222°C
Flash Point 101°C (213°F)
Refractive Index 1.497
Quantity 250 g
Beilstein 2046122
Formula Weight 164.21
Percent Purity 99%
Chemical Name or Material Benzyl propionate
EINECSNumber 204-559-3
RTECSNumber UA2537603
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.