Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl (S)-(+)-Glycidyl Ether 98.0+%, TCI America™
SDP

Catalog No. B22395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22395G 5 g
B22391G 1 g
B223925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22395G Supplier TCI America Supplier No. B22395G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16495-13-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00054428
InChI Key QNYBOILAKBSWFG-SNVBAGLBSA-N
Synonym s-+-benzyl glycidyl ether, s-benzyloxymethyl-oxirane, benzyl s-+-glycidyl ether, s-2-benzyloxy methyl oxirane, 2s-2-benzyloxy methyl oxirane, s-o-benzylglycidol, s-benzyl glycidyl ether, +-benzyl glycidyl ether, s-benzyloxymethyl oxirane
PubChem CID 146296
IUPAC Name (2S)-2-[(benzyloxy)methyl]oxirane
SMILES C(OCC1=CC=CC=C1)[C@@H]1CO1

Specifications

Color Colorless
Boiling Point 130°C
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl (S)-(+)-Glycidyl Ether
RTECSNumber TX2860020
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.