Learn More
Medchemexpress LLC Benzyl salicylate | 118-58-1 | 99.95% | 100 G
SDP
Catalog No. 5000411387
Encompass_Preferred
Benzyl salicylate is a salicylic acid benzyl ester primarily used as a fragrance additive or UV light absorber. It has been shown to inhibit Cisplatin-induced apoptosis and reactive oxygen species generation, while also attenuating Cisplatin-induced nephrotoxicity through the inhibition of the JNK/ERK/p38 signaling pathway. This product is intended for research use only.
  • Used as a fragrance additive or UV light absorber
  • Inhibits Cisplatin-induced apoptosis and ROS generation
  • Attenuates Cisplatin-induced nephrotoxicity
  • Inhibits JNK/ERK/p38 signaling pathway
  • High purity of 99.95%
Catalog No. 50-004-11387 Supplier Medchemexpress LLC Supplier No. HYB1556100G
Add to Cart
Add to Cart
Benzyl salicylate is a salicylic acid benzyl ester primarily used as a fragrance additive or UV light absorber. It has been shown to inhibit Cisplatin-induced apoptosis and reactive oxygen species generation, while also attenuating Cisplatin-induced nephrotoxicity through the inhibition of the JNK/ERK/p38 signaling pathway. This product is intended for research use only.
  • Used as a fragrance additive or UV light absorber
  • Inhibits Cisplatin-induced apoptosis and ROS generation
  • Attenuates Cisplatin-induced nephrotoxicity
  • Inhibits JNK/ERK/p38 signaling pathway
  • High purity of 99.95%