Learn More
Medchemexpress LLC Benzyl salicylate | 118-58-1 | 99.95% | 50 G
SDP
Catalog No. 5000411214
Encompass_Preferred
Benzyl salicylate is a salicylic acid benzyl ester that can be used as a fragrance additive or UV light absorber.
  • Fragrance additive
  • UV light absorber
  • Inhibits cisplatin-induced apoptosis and ROS generation, and attenuates cisplatin-induced nephrotoxicity through inhibition of JNK/ERK/p38 signaling pathway (in vitro studies)
Catalog No. 50-004-11214 Supplier Medchemexpress LLC Supplier No. HYB155650G
Benzyl salicylate is a salicylic acid benzyl ester that can be used as a fragrance additive or UV light absorber.
  • Fragrance additive
  • UV light absorber
  • Inhibits cisplatin-induced apoptosis and ROS generation, and attenuates cisplatin-induced nephrotoxicity through inhibition of JNK/ERK/p38 signaling pathway (in vitro studies)