Learn More
Medchemexpress LLC Benzyl salicylate-d4 | 1219802-40-0 | 1 MG
SDP
Catalog No. 5000411215
Encompass_Preferred
Benzyl salicylate-d4 is a deuterium-labeled form of Benzyl salicylate, which is a salicylic acid benzyl ester. It can function as a fragrance additive or a UV light absorber. This deuterium-labeled compound is valuable as a tracer for quantitation during drug development due to stable heavy isotopes. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Benzyl salicylate (HY-B1556).
  • Can be used as a fragrance additive.
  • Functions as a UV light absorber.
  • Stable heavy isotopes for tracing in drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11215 Supplier Medchemexpress LLC Supplier No. HYB1556S1MG
Benzyl salicylate-d4 is a deuterium-labeled form of Benzyl salicylate, which is a salicylic acid benzyl ester. It can function as a fragrance additive or a UV light absorber. This deuterium-labeled compound is valuable as a tracer for quantitation during drug development due to stable heavy isotopes. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Benzyl salicylate (HY-B1556).
  • Can be used as a fragrance additive.
  • Functions as a UV light absorber.
  • Stable heavy isotopes for tracing in drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.