Learn More
Medchemexpress LLC Benzyl salicylate-d4 | 1219802-40-0 | 232.27 | C14H8D4O3 | 10 MG

Supplier: Medchemexpress LLC HYB1556S10MG
Benzyl salicylate-d4 is the deuterated isotopologue of benzyl salicylate, provided as an isotope-labeled analytical standard for research applications such as mass spectrometry and tracer studies. It is intended for use as an internal standard, reference compound, or tracer in quantitative assays and method development.
- Isotope-labeled analytical standard for mass spectrometry and tracer studies.
- Molecular formula C14H8D4O3; molecular weight 232.27.
- CAS number 1219802-40-0.
- Pack size 10 MG with example stock preparation volumes for a 10 MG pack.
- Recommended storage: pure form -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
- Suitable for quantitative assays, method validation, and pathway-targeted studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.