Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyl selenocyanate | 4671-93-6 | 99.9% | 196.11 | C8H7NSe | 5 MG
SDP

Catalog No. 5000263205
Encompass_Preferred

Benzyl selenocyanate (CAS 4671-93-6) is a small-molecule chemopreventive agent and research reagent that inhibits DNA (cytosine-5)-methyltransferase (Mtase) with a reported IC50 of about 8.4 μM. It is supplied as a high-purity solid intended for preclinical and laboratory studies, with accompanying product data sheet and safety data sheet.

  • High purity: 99.85%.
  • Molecular formula: C8H7NSe.
  • Molecular weight: 196.11 g·mol⁻¹.
  • Reported biological activity: Mtase inhibitor, IC50 ≈ 8.4 μM.
  • Available in small mg-scale quantities for lab use.
  • Documentation: product data sheet and SDS provided.

Catalog No. 50-002-63205 Supplier Medchemexpress LLC Supplier No. HY1319915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl selenocyanate (CAS 4671-93-6) is a small-molecule chemopreventive agent and research reagent that inhibits DNA (cytosine-5)-methyltransferase (Mtase) with a reported IC50 of about 8.4 μM. It is supplied as a high-purity solid intended for preclinical and laboratory studies, with accompanying product data sheet and safety data sheet.

  • High purity: 99.85%.
  • Molecular formula: C8H7NSe.
  • Molecular weight: 196.11 g·mol⁻¹.
  • Reported biological activity: Mtase inhibitor, IC50 ≈ 8.4 μM.
  • Available in small mg-scale quantities for lab use.
  • Documentation: product data sheet and SDS provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.