Learn More
Medchemexpress LLC Benzyl selenocyanate | 4671-93-6 | 99.9% | C8H7NSe | 10MG
SDP
Catalog No. 5000261012
Encompass_Preferred
Benzyl selenocyanate is an organoselenium research compound provided as a high-purity solid for laboratory studies. It has been reported to exhibit chemopreventive activity and inhibit DNA methyltransferase in preclinical models. Supplied in a small research-scale vial, the material requires sealed storage away from moisture and handling with standard laboratory safety precautions.
  • High purity suitable for research applications.
  • Solid form for stable storage and handling.
  • Reported chemopreventive activity and Mtase inhibition.
  • Available in small research-scale packaging for experimental use.
  • Storage: sealed, keep at recommended temperatures and protect from moisture.
Catalog No. 50-002-61012 Supplier Medchemexpress LLC Supplier No. HY13199110MG
Benzyl selenocyanate is an organoselenium research compound provided as a high-purity solid for laboratory studies. It has been reported to exhibit chemopreventive activity and inhibit DNA methyltransferase in preclinical models. Supplied in a small research-scale vial, the material requires sealed storage away from moisture and handling with standard laboratory safety precautions.
  • High purity suitable for research applications.
  • Solid form for stable storage and handling.
  • Reported chemopreventive activity and Mtase inhibition.
  • Available in small research-scale packaging for experimental use.
  • Storage: sealed, keep at recommended temperatures and protect from moisture.