Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. B044025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B044025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B044025G Supplier TCI America Supplier No. B044025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 538-74-9
Molecular Formula C14H14S
Molecular Weight (g/mol) 214.33
MDL Number MFCD00004784
InChI Key LUFPJJNWMYZRQE-UHFFFAOYSA-N
Synonym dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
PubChem CID 10867
IUPAC Name [(benzylsulfanyl)methyl]benzene
SMILES C(SCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 131°C
Quantity 25 g
Formula Weight 214.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Sulfide
RTECSNumber DC0512500
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.