Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl sulphone, 98%, Thermo Scientific™

Catalog No. AC335030050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC335030050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC335030050 Supplier Thermo Scientific Chemicals Supplier No. 335030050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-32-6
Molecular Formula C14H14O2S
Molecular Weight (g/mol) 246.324
MDL Number MFCD00022036
InChI Key AWHNUHMUCGRKRA-UHFFFAOYSA-N
Synonym dibenzyl sulfone, benzyl sulfone, sulfonylbis methylene dibenzene, dibenzyl sulphone, dibenzylsulfone, benzyl sulfone; dibenzyl sulphone;, benzene, 1,1'-sulfonylbis methylene bis, phenyl benzylsulfonyl methane, benzene,1'-sulfonylbis methylene bis
PubChem CID 69282
IUPAC Name benzylsulfonylmethylbenzene
SMILES C1=CC=C(C=C1)CS(=O)(=O)CC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 149°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Formula Weight 246.3
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Benzyl sulphone, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.