Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Thiocyanate 99.0+%, TCI America™
SDP

Catalog No. T0198500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0198500G 500 g
T019825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0198500G Supplier TCI America Supplier No. T0198500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3012-37-1
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
MDL Number MFCD00001832
InChI Key ABNDFSOIUFLJAH-UHFFFAOYSA-N
Synonym tropeolin, thiocyanic acid, phenylmethyl ester, solvat 14, benzylthiocyanate, benzylrhodonid, thiocyanic acid, benzyl ester, alpha-thiocyanatotoluene, benzyl-thiocyanate, thiocyanic acid benzyl ester, phenylmethyl thiocyanate
PubChem CID 18170
ChEBI CHEBI:16017
IUPAC Name (benzylsulfanyl)carbonitrile
SMILES N#CSCC1=CC=CC=C1

Specifications

Melting Point 42°C
Color White
Boiling Point 256°C
Quantity 500 g
UN Number 1759
Formula Weight 149.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Thiocyanate
RTECSNumber XK8155000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.