Learn More
Cayman Chemical Benzyl2ACTAmIdo2deoxya 100mg
SDP
Catalog No. 502991164
Encompass_Preferred
An inhibitor of mucin synthesis; inhibits glucosamine labeling in Caco-2, HT-29, and T84 colon cancer cells at 2 mM; enhances 5-fluorouracil-induced cell death in Capan-1 and HPAF-II cells at 0.4, 0.6, and 0.8 mg/ml; LS 174T colon cancer cells grown in the presence of benzyl-α-GalNAc exhibit reduced liver metastasis in a mouse xenograft model compared with control LS 174T cells
Catalog No. 50-299-1164 Supplier Cayman Chemical Supplier No. 31747100 MG
An inhibitor of mucin synthesis; inhibits glucosamine labeling in Caco-2, HT-29, and T84 colon cancer cells at 2 mM; enhances 5-fluorouracil-induced cell death in Capan-1 and HPAF-II cells at 0.4, 0.6, and 0.8 mg/ml; LS 174T colon cancer cells grown in the presence of benzyl-α-GalNAc exhibit reduced liver metastasis in a mouse xenograft model compared with control LS 174T cells