Learn More
Cayman Chemical Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside, Purity: ≥95%, CAS: 3554-93-6, 50 mg
SDP
Catalog No. 502992599
Encompass_Preferred
An inhibitor of mucin synthesis; inhibits glucosamine labeling in Caco-2, HT-29, and T84 colon cancer cells at 2 mM; enhances 5-fluorouracil-induced cell death in Capan-1 and HPAF-II cells at 0.4, 0.6, and 0.8 mg/ml; LS 174T colon cancer cells grown in the presence of benzyl-α-GalNAc exhibit reduced liver metastasis in a mouse xenograft model compared with control LS 174T cells
Catalog No. 50-299-2599 Supplier Cayman Chemical Supplier No. 3174750 MG
An inhibitor of mucin synthesis; inhibits glucosamine labeling in Caco-2, HT-29, and T84 colon cancer cells at 2 mM; enhances 5-fluorouracil-induced cell death in Capan-1 and HPAF-II cells at 0.4, 0.6, and 0.8 mg/ml; LS 174T colon cancer cells grown in the presence of benzyl-α-GalNAc exhibit reduced liver metastasis in a mouse xenograft model compared with control LS 174T cells