Learn More
Medchemexpress LLC HY-106406 5mg , Benzylacyclouridine CAS:82857-69-0 Purity:>98%
SDP
Catalog No. 502261638
Encompass_Preferred
HY-106406 5mg Benzylacyclouridine CAS: 82857-69-0 Benzylacyclouridine (BAU) is a potent and specific inhibitor of uridine phosphorylase, the first enzyme in the catabolism of uridine. Benzylacyclouridine can modulate the cytotoxic side effects of 5-fluorouracil (5-FU) and its derivatives. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1638 Supplier Medchemexpress LLC Supplier No. HY1064065MG
HY-106406 5mg Benzylacyclouridine CAS: 82857-69-0 Benzylacyclouridine (BAU) is a potent and specific inhibitor of uridine phosphorylase, the first enzyme in the catabolism of uridine. Benzylacyclouridine can modulate the cytotoxic side effects of 5-fluorouracil (5-FU) and its derivatives. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.