Learn More
Medchemexpress LLC Benzylamine hydrochloride | 3287-99-8 | 98.0% | C7H10ClN | 100 G
SDP
Catalog No. 5000434119
Encompass_Preferred
Benzylamine hydrochloride, also known as Phenylmethylamine, is a chemical compound primarily used as a substrate for specific enzymatic reactions. It acts as a substrate for both benzylamine oxidase and monoamine oxidase B, making it valuable in biochemical assays and research related to enzyme function. This product is supplied as a white to off-white solid.
  • Serves as a substrate for benzylamine oxidase
  • Functions as a substrate for monoamine oxidase B
  • Appears as a white to off-white solid
  • Soluble in DMSO at 50 mg/mL (348.17 mM)
Catalog No. 50-004-34119 Supplier Medchemexpress LLC Supplier No. HY76253100G
Benzylamine hydrochloride, also known as Phenylmethylamine, is a chemical compound primarily used as a substrate for specific enzymatic reactions. It acts as a substrate for both benzylamine oxidase and monoamine oxidase B, making it valuable in biochemical assays and research related to enzyme function. This product is supplied as a white to off-white solid.
  • Serves as a substrate for benzylamine oxidase
  • Functions as a substrate for monoamine oxidase B
  • Appears as a white to off-white solid
  • Soluble in DMSO at 50 mg/mL (348.17 mM)