Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzylamine hydrochloride | 3287-99-8 | 98.0% | C7H10ClN | 250 G
SDP

Catalog No. 5000358425
Encompass_Preferred

Benzylamine hydrochloride is a chemical compound that serves as a substrate for benzylamine oxidase and monoamine oxidase B.

  • Appearance: White to off-white solid
  • Molecular weight: 143.62
  • Storage (solid): Store at room temperature, keep dry and cool
  • Storage (in solvent): -80°C for 1 year, -20°C for 6 months
  • Solubility: DMSO at 50 mg/mL
  • Target: Biochemical assay reagents

Catalog No. 50-003-58425 Supplier Medchemexpress LLC Supplier No. 250GHY76253
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzylamine hydrochloride is a chemical compound that serves as a substrate for benzylamine oxidase and monoamine oxidase B.

  • Appearance: White to off-white solid
  • Molecular weight: 143.62
  • Storage (solid): Store at room temperature, keep dry and cool
  • Storage (in solvent): -80°C for 1 year, -20°C for 6 months
  • Solubility: DMSO at 50 mg/mL
  • Target: Biochemical assay reagents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.