Learn More
Medchemexpress LLC Benzylamine hydrochloride | 3287-99-8 | MFCD00012852 | 98.0% | C7H10ClN | 500 G
SDP
Catalog No. 5000433896
Encompass_Preferred
Benzylamine hydrochloride is a chemical compound that serves as a substrate for important enzymes in biological systems. It appears as a white to off-white solid and should be stored at room temperature, kept dry and cool.
  • Substrate for benzylamine oxidase
  • Substrate for monoamine oxidase B
  • White to off-white solid appearance
  • Store at room temperature in a dry, cool place
Catalog No. 50-004-33896 Supplier Medchemexpress LLC Supplier No. HY76253500G
Benzylamine hydrochloride is a chemical compound that serves as a substrate for important enzymes in biological systems. It appears as a white to off-white solid and should be stored at room temperature, kept dry and cool.
  • Substrate for benzylamine oxidase
  • Substrate for monoamine oxidase B
  • White to off-white solid appearance
  • Store at room temperature in a dry, cool place