Learn More
AdipoGen Benzylamino-NBD
SDP
Supplier:  AdipoGen CDXB0070M250
Encompass_Preferred
Chemical. CAS 18378-20-6. Formula C13H10N4O3. MW 270.2. 4-Benzylamino-7-nitrobenzofurazan is a fluorescent probe. It exhibits absorption and emission bands that are sensitive to solvent polarity, and is used as fluorescent probes in the study of micellar and lipid bilayer structures. Shorter fluorescence lifetimes are observed with more polar solvents, making the probes suited for studies of binary solvent systems. Spectral Data lambdaEx/lambdaEm EtOH = 460/540nm.
Catalog No. 50-256-1688