Learn More
Medchemexpress LLC Benzylboronic acid pinacol ester | 87100-28-5 | 218.10 | 25 G
SDP
Catalog No. 5000449995
Encompass_Preferred
Benzylboronic acid pinacol ester is a biochemical reagent suitable for life science research. With a purity of 97.0%, it appears as a colorless to off-white solid-liquid mixture and is soluble in DMSO at 100 mg/mL. This compound is utilized in Pd(0)-mediated [11C] carbonylation reactions.
  • Biochemical reagent for life science research.
  • Utilized in Pd(0)-mediated carbonylation reactions.
  • Soluble in DMSO at 100 mg/mL.
  • Pure form storage: -20°C for 3 years.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month.
Catalog No. 50-004-49995 Supplier Medchemexpress LLC Supplier No. 25GHYW000993
Benzylboronic acid pinacol ester is a biochemical reagent suitable for life science research. With a purity of 97.0%, it appears as a colorless to off-white solid-liquid mixture and is soluble in DMSO at 100 mg/mL. This compound is utilized in Pd(0)-mediated [11C] carbonylation reactions.
  • Biochemical reagent for life science research.
  • Utilized in Pd(0)-mediated carbonylation reactions.
  • Soluble in DMSO at 100 mg/mL.
  • Pure form storage: -20°C for 3 years.
  • In solvent storage: -80°C for 6 months; -20°C for 1 month.