Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyldimethyl-n-tetradecylammonium chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog No. AAJ6346514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6346514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6346514 Supplier Thermo Scientific Chemicals Supplier No. J6346514
Add to Cart

missing translation for 'validMainframeMsg'

Phase transfer catalyst

Phase transfer catalyst

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-08-2
Molecular Formula C23H42ClN
Molecular Weight (g/mol) 368.05
MDL Number MFCD00011771
InChI Key OCBHHZMJRVXXQK-UHFFFAOYSA-M
Synonym miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
PubChem CID 8755
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Color White
Linear Formula CH3(CH2)13N(CH3)2(CH2C6H5)Cl
Quantity 25 g
UN Number UN3261
Beilstein 4062599
Merck Index 14,1059
IUPAC Name benzyl-dimethyl-tetradecylazanium;chloride
Formula Weight 368.05 (Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Benzyldimethyl-n-tetradecylammonium chloride hydrate
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 205-352-0
RTECSNumber BO7150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.