Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyldimethyldodecylammonium Chloride

Catalog No. 1338050GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-380-50GF 50 g
13-380-10GF 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 13-380-50GF Supplier Sigma Aldrich Supplier No. 1338050GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyldimethyldodecylammonium chloride, a quaternary ammonium compound (QAC), is a cationic surfactant that shows antimicrobial activity against Gram-negative bacterium Pseudomonas fluorescens. It can be used as a:corrosion inhibitor.synergist to increase the toxicity of the pesticide, indoxacarb against various pests.model for benzyltrialkylammonium salts to study the thermal degradation pathway of benzyl QACs.

Specifications

CAS 139-07-1
Molecular Formula C21H38ClN
Linear Formula CH3(CH2)11N(Cl)(CH3)2CH2C6H5
MDL Number MFCD00137276
Quantity 50 g
Synonym Benzododecinium chloride; Benzyldodecyldimethylammonium chloride
Molecular Weight (g/mol) 339.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.