Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyldimethylsilane, 96%

Catalog No. AAL0533406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0533406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0533406 Supplier Thermo Scientific Chemicals Supplier No. L0533406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1631-70-5 | C9H13Si | 149.288 g/mol

Benzyldimethylsilane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyldimethylsilane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1631-70-5
Molecular Formula C9H13Si
Molecular Weight (g/mol) 149.288
MDL Number MFCD00039792
InChI Key SKDKWDVYZOKBPL-UHFFFAOYSA-N
Synonym benzyldimethylsilane, dimethyl phenylmethyl silane, silane, dimethyl phenylmethyl, benzyldimethylsilyl, benzyl dimethyl silicon, benzene, dimethylsilyl methyl
PubChem CID 6328719
IUPAC Name benzyl(dimethyl)silicon
SMILES C[Si](C)CC1=CC=CC=C1

Specifications

Boiling Point 69°C to 71°C (15mmHg)
Quantity 5 g
Flash Point 53°C (127°F)
Refractive Index 1.5026
Density 0.949
UN Number UN1993
Beilstein 2935572
Solubility Information Insoluble in water.
Formula Weight 150.3
Percent Purity 96%
Chemical Name or Material Benzyldimethylsilane
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.