Learn More
Medchemexpress LLC Benzyldimethylstearylammonium chloride | 122-19-0 | 98.0% | 500 MG
SDP
Catalog No. 5000342019
Encompass_Preferred
Benzyldimethylstearylammonium chloride is a quaternary ammonium cationic surfactant that has been shown to be without embryotoxicity. This compound has been investigated for its effectiveness as an inhibitor for cold-rolled steel in trichloroacetic acid solutions.
  • Quaternary ammonium cationic surfactant
  • Exhibits non-embryotoxic properties
  • Effective as an inhibitor for cold-rolled steel in trichloroacetic acid solution
  • Solid appearance, white to off-white color
  • Purity of 98.0%
Catalog No. 50-003-42019 Supplier Medchemexpress LLC Supplier No. HY128443500MG
Benzyldimethylstearylammonium chloride is a quaternary ammonium cationic surfactant that has been shown to be without embryotoxicity. This compound has been investigated for its effectiveness as an inhibitor for cold-rolled steel in trichloroacetic acid solutions.
  • Quaternary ammonium cationic surfactant
  • Exhibits non-embryotoxic properties
  • Effective as an inhibitor for cold-rolled steel in trichloroacetic acid solution
  • Solid appearance, white to off-white color
  • Purity of 98.0%