Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyldimethyltetradecylammonium Chloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. A51605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51605G Supplier TCI America Supplier No. A51605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Ion association reagent for extraction photometric analysis and sensitizer in photometric determination of metals]

Chemical Identifiers

CAS 139-08-2
Molecular Formula C23H42ClN
Molecular Weight (g/mol) 368.05
MDL Number MFCD00011771
InChI Key OCBHHZMJRVXXQK-UHFFFAOYSA-M
Synonym miristalkonium chloride, n-benzyl-n,n-dimethyltetradecan-1-aminium chloride, zephiramine, benzyldimethyltetradecylammonium chloride, tetradecyldimethylbenzylammonium chloride, myristalkonium chloride, dibactol, faringets, quaternario 14b, sanibond 200lg
PubChem CID 8755
IUPAC Name benzyldimethyltetradecylazanium chloride
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Color White
Quantity 5 g
UN Number 2923
Formula Weight 368.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyldimethyltetradecylammonium Chloride Hydrate [Ion association reagent for extraction photometri
EINECSNumber (3)-2694
RTECSNumber BO7150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.