Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyldodecyldimethylammonium Bromide

Catalog No. 1337350GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-373-50GF 50 g
13-373-10GF 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 13-373-50GF Supplier Sigma Aldrich Supplier No. 1337350GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyldodecyldimethylammonium Bromide

Specifications

CAS 7281-04-1
Molecular Formula C21H38BrN
Linear Formula CH3(CH2)11N(Br)(CH3)2CH2C6H5
MDL Number MFCD00011768
Quantity 50 g
Synonym Benzyldimethyldodecylammonium bromide
Molecular Weight (g/mol) 384.44 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.