Learn More
Medchemexpress LLC Benzylideneacetone | 122-57-6 | 99.7% | 146.19 | 50 G
SDP
Catalog No. 5000367036
Encompass_Preferred
Benzylideneacetone is an orally active antibiotic, tyrosinase inhibitor, phospholipase A2 inhibitor, and immunosuppressant. It exhibits antibacterial activity against certain gram-negative plant-pathogenic bacteria and can be used in the synthesis of chemicals and drugs, as well as a flavoring additive for some foods.
  • Orally active antibiotic
  • Tyrosinase inhibitor
  • Phospholipase A2 inhibitor
  • Immunosuppressant
  • Antibacterial activity against gram-negative plant-pathogenic bacteria
  • Used in synthesis of chemicals and drugs
  • Functions as a flavoring additive
Catalog No. 50-003-67036 Supplier Medchemexpress LLC Supplier No. HYW01259550G
Benzylideneacetone is an orally active antibiotic, tyrosinase inhibitor, phospholipase A2 inhibitor, and immunosuppressant. It exhibits antibacterial activity against certain gram-negative plant-pathogenic bacteria and can be used in the synthesis of chemicals and drugs, as well as a flavoring additive for some foods.
  • Orally active antibiotic
  • Tyrosinase inhibitor
  • Phospholipase A2 inhibitor
  • Immunosuppressant
  • Antibacterial activity against gram-negative plant-pathogenic bacteria
  • Used in synthesis of chemicals and drugs
  • Functions as a flavoring additive