Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylideneaniline 98.0+%, TCI America™
SDP

Catalog No. B103425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B103425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B103425G Supplier TCI America Supplier No. B103425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 538-51-2
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.24
MDL Number MFCD00003027
InChI Key UVEWQKMPXAHFST-UHFFFAOYSA-N
Synonym n-benzylideneaniline, benzalaniline, benzylideneaniline, e-n-benzylideneaniline, n-benzylidenaniline, n-benzalaniline, benzaldehyde anil, aniline, n-benzylidene, benzenamine, n-phenylmethylene, n-phenylmethylene benzenamine
PubChem CID 10858
IUPAC Name N,1-diphenylmethanimine
SMILES C(=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52°C
Color Yellow
Boiling Point 300°C
Quantity 25 g
Formula Weight 181.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylideneaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.