Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Benzylmethylphenylsilylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. B1721100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1721100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1721100MG Supplier TCI America Supplier No. B1721100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95349-35-2
Molecular Formula C16H18O2Si
Molecular Weight (g/mol) 270.403
MDL Number MFCD00191393
InChI Key JIPQZCPYHVQEIZ-UHFFFAOYSA-N
PubChem CID 13698775
IUPAC Name 2-(benzyl-methyl-phenylsilyl)acetic acid
SMILES C[Si](CC1=CC=CC=C1)(CC(=O)O)C2=CC=CC=C2

Specifications

Melting Point 70°C
Color White
Quantity 100 mg
Formula Weight 270.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Benzylmethylphenylsilylacetic Acid [for e.e. Determination by NMR]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.