Learn More
Medchemexpress LLC Benzyloxy-C5-PEG1 100mg
SDP
Catalog No. 5000205759
Encompass_Preferred
Benzyloxy-C5-PEG1 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05759 Supplier Medchemexpress LLC Supplier No. HYW096134100MG
Benzyloxy-C5-PEG1 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]