Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyloxycarbonyl-alanyl-glutamyl-valyl-aspartic acid fluoromethyl ketone | 1135688-47-9 | MFCD03453601 | >=95.0% | 610.63 | C28H39FN4O10 | 5MG
SDP

Catalog No. 5000237759
Encompass_Preferred

Z-AEVD-FMK is a cell-permeable, fluoromethyl ketone-derivatized peptide that irreversibly inhibits caspase-10 and related caspases. It prevents Bid activation and downstream caspase cascade events and is supplied as a crystalline solid for laboratory research use only.

  • Irreversible caspase-10 inhibitor with FMK warhead.
  • Cell-permeable peptide suitable for intracellular studies.
  • CAS number 1135688-47-9; molecular formula C28H39FN4O10.
  • Molecular weight approximately 610.6 Da.
  • Purity ≥95% (reported by distributor).
  • Soluble in DMSO at ~10 mg/mL; supplied as a crystalline solid.
  • For research use only; not for human or veterinary applications.

Catalog No. 50-002-37759 Supplier Medchemexpress LLC Supplier No. HY1253055MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-AEVD-FMK is a cell-permeable, fluoromethyl ketone-derivatized peptide that irreversibly inhibits caspase-10 and related caspases. It prevents Bid activation and downstream caspase cascade events and is supplied as a crystalline solid for laboratory research use only.

  • Irreversible caspase-10 inhibitor with FMK warhead.
  • Cell-permeable peptide suitable for intracellular studies.
  • CAS number 1135688-47-9; molecular formula C28H39FN4O10.
  • Molecular weight approximately 610.6 Da.
  • Purity ≥95% (reported by distributor).
  • Soluble in DMSO at ~10 mg/mL; supplied as a crystalline solid.
  • For research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.