Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyloxycarbonyl-L-leucyl-L-leucyl-L-glutamic acid 7-amido-4-methylcoumarin | 348086-66-8 | MFCD01074989 | 99.5% | 664.75 g·mol⁻¹ | C35H44N4O9 | 50MG
SDP

Catalog No. 5000238534
Encompass_Preferred

Z-Leu-Leu-Glu-AMC is a fluorogenic peptide substrate for measuring postacidic-like (peptidylglutamyl-peptide hydrolyzing, PGPH) activity of the 20S proteasome. Proteolytic cleavage liberates the fluorescent 7-amino-4-methylcoumarin (AMC) reporter, enabling quantitative proteasome activity assays for research use.

  • Fluorogenic substrate that releases 7-amino-4-methylcoumarin (AMC) upon cleavage.
  • Designed to measure postacidic-like (PGPH) activity of the 20S proteasome.
  • Reported purity 99.5%.
  • Chemical formula C35H44N4O9.
  • Molecular weight 664.75 g·mol⁻¹.
  • White to off-white solid appearance.
  • Available in multiple pack sizes including 50 mg.
  • Storage: powder stable at -80°C for long-term and -20°C for short-term when sealed and protected from moisture.

Catalog No. 50-002-38534 Supplier Medchemexpress LLC Supplier No. HY12305350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Leu-Leu-Glu-AMC is a fluorogenic peptide substrate for measuring postacidic-like (peptidylglutamyl-peptide hydrolyzing, PGPH) activity of the 20S proteasome. Proteolytic cleavage liberates the fluorescent 7-amino-4-methylcoumarin (AMC) reporter, enabling quantitative proteasome activity assays for research use.

  • Fluorogenic substrate that releases 7-amino-4-methylcoumarin (AMC) upon cleavage.
  • Designed to measure postacidic-like (PGPH) activity of the 20S proteasome.
  • Reported purity 99.5%.
  • Chemical formula C35H44N4O9.
  • Molecular weight 664.75 g·mol⁻¹.
  • White to off-white solid appearance.
  • Available in multiple pack sizes including 50 mg.
  • Storage: powder stable at -80°C for long-term and -20°C for short-term when sealed and protected from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.