Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyloxycarbonyl-L-valylglycine | 2790-84-3 | >98.0% | 308.33 g/mol | C15H20N2O5 | 10 G
SDP

Catalog No. 5000210564
Encompass_Preferred

Z-Val-Gly-OH is the N-benzyloxycarbonyl (Cbz, Z) protected dipeptide of valine and glycine supplied as a research reagent and synthetic intermediate. It is commonly used in peptide synthesis and biochemical studies.

  • Cbz-protected dipeptide of valine and glycine
  • Purity greater than 98.0%
  • Molecular weight 308.33 g/mol
  • Chemical formula C15H20N2O5
  • CAS number 2790-84-3
  • Available pack sizes: 1 g, 10 g, 25 g

Catalog No. 50-002-10564 Supplier Medchemexpress LLC Supplier No. HYW04883010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Val-Gly-OH is the N-benzyloxycarbonyl (Cbz, Z) protected dipeptide of valine and glycine supplied as a research reagent and synthetic intermediate. It is commonly used in peptide synthesis and biochemical studies.

  • Cbz-protected dipeptide of valine and glycine
  • Purity greater than 98.0%
  • Molecular weight 308.33 g/mol
  • Chemical formula C15H20N2O5
  • CAS number 2790-84-3
  • Available pack sizes: 1 g, 10 g, 25 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.