Learn More
Apexbio Technology LLC Benzylpenicillin potassium 113-98-4 500mg
SDP
Catalog No. 5000094548
Encompass_Preferred
Benzylpenicillin potassium (113-98-4) is a beta-lactam antibiotic targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall biosynthesis by interfering with peptidoglycan cross-linking thereby reducing bacterial stability and inducing cell lysis Benzylpenicillin potassium exerts its biological activity primarily through inhibition of PBPs To date specific IC50 concentrations of Benzylpenicillin potassium vary according to bacterial strains experimental conditions and assay methodologies employed in antimicrobial susceptibility studies Based on these pharmacological properties Benzylpenicillin potassium holds research potential in investigating bacterial infections associated with blood cardiac tissue pulmonary structures joints and reproductive organs
Catalog No. 50-000-94548 Supplier Apexbio Technology LLC Supplier No. C7124500
Benzylpenicillin potassium (113-98-4) is a beta-lactam antibiotic targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall biosynthesis by interfering with peptidoglycan cross-linking thereby reducing bacterial stability and inducing cell lysis Benzylpenicillin potassium exerts its biological activity primarily through inhibition of PBPs To date specific IC50 concentrations of Benzylpenicillin potassium vary according to bacterial strains experimental conditions and assay methodologies employed in antimicrobial susceptibility studies Based on these pharmacological properties Benzylpenicillin potassium holds research potential in investigating bacterial infections associated with blood cardiac tissue pulmonary structures joints and reproductive organs