Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Benzylphenethylamine | 38235-77-7 | 99.89% | 211.30 | 100 G
SDP

Catalog No. 5000403665
Encompass_Preferred

(+)-Benzylphenethylamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Purity: 99.89%
  • Molecular weight: 211.30
  • CAS number: 38235-77-7
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage: 4°C, protect from light

Catalog No. 50-004-03665 Supplier Medchemexpress LLC Supplier No. HY41390100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Benzylphenethylamine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Purity: 99.89%
  • Molecular weight: 211.30
  • CAS number: 38235-77-7
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage: 4°C, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.