Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzylthiouracil (6-benzyl-2-thiouracil) | 6336-50-1 | 98.7% | 218.28 g/mol | C11H10N2OS | 250 MG
SDP

Catalog No. 5000209664
Encompass_Preferred

Benzylthiouracil is an antithyroid active pharmaceutical ingredient used in research on hyperthyroidism and TSH receptor inhibition. Supplied for laboratory research use only.

  • Antithyroid active pharmaceutical ingredient used in research.
  • CAS number 6336-50-1.
  • Purity 98.7% (typical).
  • Molecular formula C11H10N2OS, molecular weight 218.28 g/mol.
  • Available as solid (25 mg-250 mg) and 10 mM solution in DMSO.

Catalog No. 50-002-09664 Supplier Medchemexpress LLC Supplier No. 250MGHYW069177
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzylthiouracil is an antithyroid active pharmaceutical ingredient used in research on hyperthyroidism and TSH receptor inhibition. Supplied for laboratory research use only.

  • Antithyroid active pharmaceutical ingredient used in research.
  • CAS number 6336-50-1.
  • Purity 98.7% (typical).
  • Molecular formula C11H10N2OS, molecular weight 218.28 g/mol.
  • Available as solid (25 mg-250 mg) and 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.