Learn More
Medchemexpress LLC Benzyltriethylammonium chloride | 56-37-1 | MFCD00011824 | ≥98.0% | 227.77 g·mol⁻¹ | C13H22ClN | 1000g
SDP
Catalog No. 5000269343
Encompass_Preferred
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69343 Supplier Medchemexpress LLC Supplier No. HYY06671000G
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes