Learn More
Medchemexpress LLC Benzyltriethylammonium chloride | 56-37-1 | MFCD00011824 | ≥98.0% | 227.77 g/mol | C13H22ClN | 100g
SDP
Catalog No. 5000268959
Encompass_Preferred
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-68959 Supplier Medchemexpress LLC Supplier No. HYY0667100G
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes