Learn More
Medchemexpress LLC Benzyltriethylammonium chloride | 56-37-1 | MFCD00011824 | ≥98.0% | 227.77 g/mol | C13H22ClN | 500g
SDP
Catalog No. 5000269328
Encompass_Preferred
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69328 Supplier Medchemexpress LLC Supplier No. HYY0667500G
Benzyltriethylammonium chloride is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Benzyltriethylammonium chloride has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes