Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltrimethylammonium bromide, 98+%

Catalog No. AAA1684014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1684014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1684014 Supplier Thermo Scientific Chemicals Supplier No. A1684014

missing translation for 'validMainframeMsg'

CAS: 5350-41-4 | C10H16BrN | 230.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5350-41-4
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.149
MDL Number MFCD00011780
InChI Key UUZYBYIOAZTMGC-UHFFFAOYSA-M
Synonym benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btm
PubChem CID 21449
IUPAC Name benzyl(trimethyl)azanium;bromide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point 228°C to 232°C
Quantity 25 g
Beilstein 3917251
Sensitivity Hygroscopic
Formula Weight 230.15
Percent Purity ≥98%
Chemical Name or Material Benzyltrimethylammonium bromide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 226-320-2
RTECSNumber BO8392000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.