Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyltrimethylammonium Hydroxide Solution

Catalog No. 246034500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-603-4500G 500 g
24-603-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 24-603-4500G Supplier Sigma Aldrich Supplier No. 246034500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyltrimethylammonium hydroxide can be used:As a base in the synthesis of activated ynesulfonamides and tertiary enesulfonamides.In the synthesis of 3-indolyl-3-hydroxy oxindoles by treating isatin with indole.As a precursor for the preparation of quaternary ammonium acetate, benzyltrimethylammonium acetate (NBz111AcO). NBz111AcO solution in DMSO can be used to dissolve cellulose.

Specifications

CAS 100-85-6
Density 1.059 g/mL at 25°C
Molecular Formula C10H17NO
Linear Formula C6H5CH2N(OH)(CH3)3
Refractive Index n20/D 1.43
Quantity 500 g
MDL Number MFCD00008281
Molecular Weight (g/mol) 167.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.