Learn More
Medchemexpress LLC Benzyltrimethylammonium tribromide | 111865-47-5 | MFCD00134423 | 98.0% | 389.95 g/mol | C10H16Br3N | 1 KG
SDP
Catalog No. 5000299687
Encompass_Preferred
Benzyltrimethylammonium tribromide is a quaternary ammonium tribromide reagent supplied as a solid for research use in organic synthesis and biochemical assays. It serves as a brominating and oxidizing agent for laboratory-scale bromination and oxidative transformations, and is provided with purity and storage guidance for reliable performance in experimental workflows.
  • High purity (98.0%) suitable for research applications.
  • Solid form for convenient storage and handling.
  • Available in multiple package sizes, including 1 kg for larger-scale use.
  • Effective brominating reagent for a variety of organic transformations.
  • Store sealed and protected from moisture to maintain stability.
Catalog No. 50-002-99687 Supplier Medchemexpress LLC Supplier No. HYW0184541KG
Benzyltrimethylammonium tribromide is a quaternary ammonium tribromide reagent supplied as a solid for research use in organic synthesis and biochemical assays. It serves as a brominating and oxidizing agent for laboratory-scale bromination and oxidative transformations, and is provided with purity and storage guidance for reliable performance in experimental workflows.
  • High purity (98.0%) suitable for research applications.
  • Solid form for convenient storage and handling.
  • Available in multiple package sizes, including 1 kg for larger-scale use.
  • Effective brominating reagent for a variety of organic transformations.
  • Store sealed and protected from moisture to maintain stability.