Learn More
Medchemexpress LLC Benzyltrimethylammonium tribromide | 111865-47-5 | MFCD00134423 | ≥98.0% | 389.95 g/mol | C10H16Br3N | 100 G
SDP
Catalog No. 5000301633
Encompass_Preferred
Benzyltrimethylammonium tribromide is a quaternary ammonium tribromide reagent used to supply controlled bromine for oxidation, cyclization, and other bromination reactions in organic synthesis. Supplied for research use, the compound is typically provided as a high-purity solid suitable for laboratory handling and storage.
  • Provides controlled bromination for oxidation and cyclization reactions.
  • High purity suitable for research applications.
  • Solid form is stable and easy to weigh and handle.
  • Dissolves readily in common polar solvents such as DMSO.
  • Available in laboratory-scale package sizes for synthetic workflows.
Catalog No. 50-003-01633 Supplier Medchemexpress LLC Supplier No. HYW018454100G
Benzyltrimethylammonium tribromide is a quaternary ammonium tribromide reagent used to supply controlled bromine for oxidation, cyclization, and other bromination reactions in organic synthesis. Supplied for research use, the compound is typically provided as a high-purity solid suitable for laboratory handling and storage.
  • Provides controlled bromination for oxidation and cyclization reactions.
  • High purity suitable for research applications.
  • Solid form is stable and easy to weigh and handle.
  • Dissolves readily in common polar solvents such as DMSO.
  • Available in laboratory-scale package sizes for synthetic workflows.