Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzyltrimethylammonium tribromide | 111865-47-5 | MFCD00134423 | 98.0% | 389.95 g/mol | C10H16Br3N | 25 G
SDP

Catalog No. 5000303104
Encompass_Preferred

Benzyltrimethylammonium tribromide is a quaternary ammonium brominating reagent (CAS 111865-47-5) used in organic synthesis for selective bromination reactions. Supplied as a yellow-orange solid for research use, it provides high purity and convenient solubility in polar aprotic solvents when prepared correctly. Handle with care due to reactive bromine content and solvent hygroscopicity.

  • High purity (98.0%).
  • Yellow-orange solid for easy identification.
  • Molecular formula C10H16Br3N; molecular weight 389.95 g/mol.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Suitable as a brominating reagent in organic synthesis.
  • Hygroscopic solvent effects; use freshly opened DMSO for best results.
  • Available in 25 g laboratory packaging.

Catalog No. 50-003-03104 Supplier Medchemexpress LLC Supplier No. 25GHYW018454
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyltrimethylammonium tribromide is a quaternary ammonium brominating reagent (CAS 111865-47-5) used in organic synthesis for selective bromination reactions. Supplied as a yellow-orange solid for research use, it provides high purity and convenient solubility in polar aprotic solvents when prepared correctly. Handle with care due to reactive bromine content and solvent hygroscopicity.

  • High purity (98.0%).
  • Yellow-orange solid for easy identification.
  • Molecular formula C10H16Br3N; molecular weight 389.95 g/mol.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Suitable as a brominating reagent in organic synthesis.
  • Hygroscopic solvent effects; use freshly opened DMSO for best results.
  • Available in 25 g laboratory packaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.