Learn More
Medchemexpress LLC Benzyltrimethylammonium tribromide | 111865-47-5 | MFCD00134423 | 98.0% | 389.95 g/mol | C10H16Br3N | 500 G
SDP
Catalog No. 5000299714
Encompass_Preferred
Benzyltrimethylammonium tribromide (CAS 111865-47-5) is a quaternary ammonium tribromide reagent used as a controlled source of electrophilic bromine in organic synthesis and biochemical assays. Supplied as a solid with high purity, it is suitable for laboratory research, oxidation, and bromination reactions.
  • High purity (≥98.0%).
  • Solid crystalline form for easy weighing and handling.
  • Provides controlled electrophilic bromine for oxidation and bromination.
  • Suitable for organic synthesis and biochemical assay applications.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-002-99714 Supplier Medchemexpress LLC Supplier No. HYW018454500G
Benzyltrimethylammonium tribromide (CAS 111865-47-5) is a quaternary ammonium tribromide reagent used as a controlled source of electrophilic bromine in organic synthesis and biochemical assays. Supplied as a solid with high purity, it is suitable for laboratory research, oxidation, and bromination reactions.
  • High purity (≥98.0%).
  • Solid crystalline form for easy weighing and handling.
  • Provides controlled electrophilic bromine for oxidation and bromination.
  • Suitable for organic synthesis and biochemical assay applications.
  • Available in multiple pack sizes for scale flexibility.